Learn More
Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide, Thermo Scientific Chemicals
Description
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.
Solubility
Slightly miscible with water.
Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkali metals, alkaline earth metals, amides, amines and powdered metals.

Specifications
Specifications
Melting Point | °C to 119°C |
Density | 1.398 |
Boiling Point | 70°C to 71°C |
Flash Point | -2°C (28°F) |
Assay Percent Range | 99% |
Linear Formula | CH2=CHCH2Br |
Refractive Index | 1.47 |
Quantity | 250 g |
UN Number | UN1099 |
Sensitivity | Light sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253303 | Certificate of Analysis | 02/12/2024 | A11766.30,A11766.0B |
10247926 | Certificate of Analysis | 06/09/2023 | A11766.30,A11766.0B |
10238020 | Certificate of Analysis | 10/01/2023 | A11766.30,A11766.0B |
10244671 | Certificate of Analysis | 18/11/2022 | A11766.30,A11766.0B |
10223724 | Certificate of Analysis | 28/04/2022 | A11766.30,A11766.0B |
Safety and Handling





- Allyl bromide, stabilized with 300-1000ppm Propylene oxide
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H400-Very toxic to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Allyl bromide, Propylene oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.