Learn More
Invitrogen™ FirstChoice™ RLM-RACE Kit
Designed to amplify cDNA only from full-length, capped mRNA, usually producing a single band after PCR
Brand: Invitrogen™ AM1700M
875.52 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
918.08 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate GAPDH-M, and 0.5M EDTA should be stored at –20°C. Nuclease-free water may be stored at any temperature.
Requires : SuperTaq Thermostable Taq DNA Polymerase sold separately. For optimal amplification of fragments ≥1 kb, use SuperTaq-Plus.
This item is not returnable. View return policy
Description
The FirstChoice RLM-RACE Kit is a major improvement over the basic rapid amplification of cDNA ends (RACE) protocol. The RLM-RACE procedure selects only full-length mRNA—no rRNA, tRNA or degraded RNA—and facilitates the cloning of sequences from the 5' ends of messages.
- From RNA to PCR product in less than a day
- Yields single, specific product from rare transcripts
- Selects 5' and/or 3' ends of true messages
- Efficient—
- Al enzymatic reactions are optimized to ensure detection of even rarest mRNA
- Final product size: 7kb or less
- Optimal reaction temperature: 42°C
- Sample: RNA
- Rapid amplification of cDNA ends (5'-RACE) is polymerase chain reaction-based technique developed to facilitate cloning of sequence from the 5'-ends of messages
- Improvement to basic RACE protocol
- Selects full-length mRNA—no rRNA, tRNA or degraded mRNA
Optimal reaction temperature: 42°C
Cloning, PCR and Real-Time PCR, Reverse Transcription, cDNA Libraries and Library Construction
Order Info
Shipping Condition: Dry ice

Specifications
RLM-RACE Kit | |
Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate™ GAPDH-M, and 0.5 M EDTA should be stored at –20°C. Nuclease-free water may be stored at any temperature. | |
42°C | |
Kit | |
cDNA Libraries & Library Construction |
Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate GAPDH-M, and 0.5 M EDTA should be stored at -20°C. Nuclease-free water may be stored at any temperature and manual | |
cDNA | |
M-MLV | |
Ambion™, FirstChoice™ | |
1 Kit |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.