Learn More
Ammonium hydroxide, 28% NH3, Thermo Scientific Chemicals
Description
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens′ reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.
Solubility
Miscible with water.
Notes
Incompatible with copper, iron and zinc.

Specifications
Specifications
pH | 12 |
Quantity | 500 mL |
Linear Formula | NH4OH |
UN Number | UN2672 |
Merck Index | 14,494 |
Solubility Information | Miscible with water. |
Formula Weight | 35.05 |
Odor | Ammonia-like |
Density | 0.9 |
Assay | 28% NH3 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z23K012 | Certificate of Analysis | 06/01/2025 | L13168.AP,L13168.AU,L13168.0F |
W17K051 | Certificate of Analysis | 25/09/2024 | L13168.AP,L13168.AU,L13168.0F |
P01K039 | Certificate of Analysis | 31/03/2024 | L13168.AP,L13168.AU,L13168.0F |
10239962 | Certificate of Analysis | 20/02/2024 | L13168.AP,L13168.AU,L13168.0F |
P21I035 | Certificate of Analysis | 20/02/2024 | L13168.AP,L13168.AU,L13168.0F |
Safety and Handling



- Ammonium hydroxide, 28% NH3
- Danger
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308-IF exposed or concerned:.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.