missing translation for 'onlineSavingsMsg'
Learn More
Ammonium thiocyanate solution, Volumetric, 0.1 M NH4SCN (0.1N), Honeywell Fluka Product Code.: 15654780

Ammonium thiocyanate solution, Volumetric, 0.1 M NH4SCN (0.1N), Honeywell Fluka

Product Code. 15654780 Shop All Honeywell Fluka Products
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15654780

Brand: Honeywell-Fluka 350361L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Volumetric, 0.1 M NH4SCN (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
CAS 1762-95-4, 7732-18-5
CAS Min % ≥99.0000%
CAS Max % ≤1.0000%
Quantity 1 L
Molecular Formula CH4N2S
Linear Formula NH4SCN
MDL Number MFCD00011428
Beilstein 3595135
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES C(#N)[S-].[NH4+]
IUPAC Name azanium;thiocyanate
Molecular Weight (g/mol) 76.117
PubChem CID 15666
Formula Weight 76.12
Concentration or Composition (by Analyte or Components) 0.1 M NH4SCN (0.1N)
Packaging Glass Bottle
Chemical Name or Material Ammonium thiocyanate solution, Volumetric, 0.1 M NH4SCN (0.1N)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium thiocyanate solution, Volumetric, 0.1 M NH4SCN (0.1N), Honeywell Fluka >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.