missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Analog SBB Aqua Plus Boiling Bath

Provides steady and continuous 100°C operation. Grant Instruments™ Analog SSB Aqua Plus Boiling Bath will not boil violently, spit, or produce excessive steam.

Brand:  Grant Instruments™ SBB AQUA 26 PLUS

1386.26 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 11466437

  • 1756.00 € / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Adjustable energy regulator provides steady boiling
  • Constant level device maintains liquid level
  • Fixed thermal cut-out - independent safety feature
  • Simple, intuitive controls - quick and easy to set temperature
  • Robust and reliable design to withstand everyday wear and tear
  • Choice of sizes to suit individual applications
  • Non-drip polycarbonate lid included as standard
  • 3 year warranty
TRUSTED_SUSTAINABILITY
Specifications

Specifications

26 L
2000 w (230V), 1350 w (120V)
570 x 385 x 300 mm
100°C
3 Years
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Grant Instruments™ Analog SBB Aqua Plus Boiling Bath > Capacity: 26L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.