missing translation for 'onlineSavingsMsg'
Learn More
Ansell ActivArmr™ 80-400 Crinkle Latex-Coated Insulated Gloves Product Code.: 11716683
Add to basket

Ansell ActivArmr™ 80-400 Crinkle Latex-Coated Insulated Gloves

Product Code. 11716683 Shop All Ansell Products
9
Click to view available options
Size:
9
This item is not returnable. View return policy

Product Code. 11716683

Brand: Ansell 80400090

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Thermal acrylic liner combined with naturally insulating latex coating provides protection from temperatures down to -30°C (-22°F) for short periods of time.

  • A general purpose glove designed to provide insulation from cold conditions
  • Thermal terry loop acrylic liner with a naturally insulating latex coating
  • Crinkle latex coating increases comfort and grip capabilities
  • Bright yellow color enhances visibility in low or poor light conditions
  • Protection from temperatures as low as 15°C (5°F) for extended periods of time, and -30°C (-22°F) for short-term contact
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Coated Area Palm, Fingers
Coating Material Crinkled latex
Color Hi-vis Yellow
Cut Resistance Rating ANSI A3
Exterior Finish Fully Textured
Hand Specific
Size 9
Temperature (English) Max. 480°F (≤15 sec.)
Temperature (Metric) Max 250°C (≤15 sec.)
For Use With (Application) Construction, Shipping/Receiving, and Outdoor Winter Use
Glove Cuff Style Knit
Liner Material Thermal Acrylic
Series 80-400
Temperature (English) Min 5°F (Extended), -22°F (Intermittent)
Temperature (Metric) Min. 15°C (Extended), -30°C (Intermittent)
Yarn Gauge 7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell ActivArmr™ 80-400 Crinkle Latex-Coated Insulated Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.