missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Brand:  Fisher Chemical K/2110/15

69.83 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.



 View more versions of this product

Product Code. 10199511

  • 109.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Solvent for samples with high hydrocarbon content
  • Used in conjunction with Aqualine™titrant for two component volumetric Karl Fischer analysis
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqualine™ Solvent CM
Pyridine free
67-66-3, 288-32-4, 7446-09-5
Liquid
81734
Karl Fischer Reagent
1 L
Amber glass bottle
Pass Test
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical™ > 1L, Amber glass bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.