missing translation for 'onlineSavingsMsg'
Learn More
Aristolochic acid I, 96%, Thermo Scientific Chemicals Product Code.: 10420501

Aristolochic acid I, 96%, Thermo Scientific Chemicals

Product Code. 10420501
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10420501

Brand: Thermo Scientific Acros 226091000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313-67-7
Molecular Formula C17H11NO7
Molecular Weight (g/mol) 341.27
MDL Number MFCD00004996
InChI Key BBFQZRXNYIEMAW-UHFFFAOYSA-N
Synonym aristolochic acid, aristolochic acid a, aristolochic acid i, aristolochin, tardolyt, birthwort, aristolochic acid-i, aristolochine, aristolochiazaeure, unii-94218wfp5tShow MoreShow Less
PubChem CID 2236
ChEBI CHEBI:2825
SMILES COC1=CC=CC2=C3C(=C(C=C21)[N+](=O)[O-])C(=CC4=C3OCO4)C(=O)O

Specifications

Melting Point 269°C to 270°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (typically I+ II isomers) (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 19, V,7, 611
Merck Index 15, 777
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform,ether,acetone,,acetic acid,aniline,alkalies,practically insoluble in benzene,carbon disulfide
Formula Weight 341.27
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Aristolochic acid, Mixture of I and II
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462531 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number226091000,226095000
Lot NumberA0431083 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number226091000,226095000
Lot NumberA0401041 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number226091000,226095000
Lot NumberA0407297 Certificate TypeCertificate of Analysis Date20/10/2021 Catalog Number226091000,226095000
compliance-icons compliance-icons
Product Identifier
  • Aristolochic acid, mixture of I and II
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1A
Hazard Statement
  • H301-Toxic if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aristolochic acid I, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.