Learn More
Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
For reduced non specific sample interaction and increased analyte recovery
66.65 € - 159.00 €
Spezifikation
For Use With (Application) Gas chromatography
Product Type Autosampler Syringe
Quantity Ea.
Mehr Spezifikationen ansehen
Products 20
Produktcode Marke Needle Gauge Diameter (English) Outer Needle Length (English) Needle Model Preis Menge & Verfügbarkeit  
Produktcode Marke Needle Gauge Diameter (English) Outer Needle Length (English) Needle Model Preis Menge & Verfügbarkeit  
13189015
missing translation for 'viewDocuments'
Trajan™
002865
26 G 0.01 in. 2.2 in. 10F-RTC/RSH-5.7/0.47C
66.65 €
Each
13186105
missing translation for 'viewDocuments'
Trajan™
001861
23 G 0.02 in. 2.2 in. 5F-RTC/RSH-5.7/0.63C
87.55 €
Each
13199195
missing translation for 'viewDocuments'
Trajan™
002866
26 G 0.01 in. 2.2 in. 10F-RTC/RSH-GT-5.7/0.47C
85.10 €
Each
13179015
missing translation for 'viewDocuments'
Trajan™
002863
23 G 0.02 in. 3.3 in. 10F-RTC/RSH-8.5/0.63C
87.55 €
Each
13109205
missing translation for 'viewDocuments'
Trajan™
002868
26 G 0.01 in. 3.3 in. 10F-RTC/RSH-GT-8.5/0.47C
93.20 €
Each
13109215
missing translation for 'viewDocuments'
Trajan™
004864
23 G 0.02 in. 3.3 in. 50F-RTC/RSH-GT-8.5/0.63C
143.00 €
Each
13106115
missing translation for 'viewDocuments'
Trajan™
001865
26 G 0.01 in. 2.2 in. 5F-RTC/RSH-5.7/0.47C
87.55 €
Each
13169015
missing translation for 'viewDocuments'
Trajan™
002861
23 G 0.02 in. 2.2 in. 10F-RTC/RSH-5.7/0.63C
66.65 €
Each
13196105
missing translation for 'viewDocuments'
Trajan™
001863
23 G 0.02 in. 3.3 in. 5F-RTC/RSH-8.5/0.63C
119.00 €
Each
13129215
missing translation for 'viewDocuments'
Trajan™
004868
26 G 0.01 in. 3.3 in. 50F-RTC/RSH-GT-8.5/0.47C
143.00 €
Each
13199205
missing translation for 'viewDocuments'
Trajan™
004862
23 G 0.02 in. 2.2 in. 50F-RTC/RSH-GT-5.7/0.63C
115.00 €
Each
13179195
missing translation for 'viewDocuments'
Trajan™
002862
23 G 0.02 in. 2.2 in. 10F-RTC/RSH-GT-5.7/0.63C
85.10 €
Each
13189205
missing translation for 'viewDocuments'
Trajan™
003868
26 G 0.01 in. 3.3 in. 25FRTC/RSH-GT-8.5/0.47C
141.00 €
Each
13179205
missing translation for 'viewDocuments'
Trajan™
003866
26 G 0.01 in. 2.2 in. 25F-RTC/RSH-GT-5.7/0.47C
116.00 €
Each
13119215
missing translation for 'viewDocuments'
Trajan™
004866
26 G 0.01 in. 2.2 in. 50F-RTC/RSH-GT-5.7/0.47C
115.00 €
Each
13189195
missing translation for 'viewDocuments'
Trajan™
002864
23 G 0.02 in. 3.3 in. 10F-RTC/RSH-GT-8.5/0.63C
93.20 €
Each
13159205
missing translation for 'viewDocuments'
Trajan™
003862
23 G 0.02 in. 2.2 in. 25F-RTC/RSH-GT-5.7/0.63C
116.00 €
Each
13169205
missing translation for 'viewDocuments'
Trajan™
003864
23 G 0.02 in. 3.3 in. 25F-RTC/RSH-GT-8.5/0.63C
141.00 €
Each
13199015
missing translation for 'viewDocuments'
Trajan™
002867
26 G 0.01 in. 3.3 in. 10F-RTC/RSH-8.5/0.47C
87.55 €
Each
12977506
missing translation for 'viewDocuments'
Trajan™
003988
26 G 0.018 in. 1.9 in. 25R-C/T-GT-0.47
159.00 €
Each
Beschreibung
Beschreibung
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Spezifikation
Spezifikation
Gas chromatography
Autosampler Syringe
Ea.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate