missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Avidin, NeutrAvidin™, FITC conjugate

Avidin, NeutrAvidin™, FITC conjugate

Brand:  Molecular Probes™ A2662

212.04 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10013522

  • 279.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FITC (Fluorescein)
Store in freezer (-5 to -30°C) and protect from light.
NeutrAvidin™
1 mg
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2735074 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog NumberA2662
Lot Number2577232 Certificate TypeCertificate of Analysis Date20/12/2022 Catalog NumberA2662
Lot Number2456874 Certificate TypeCertificate of Analysis Date20/01/2022 Catalog NumberA2662
Lot Number2306802 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog NumberA2662
Lot Number2214537 Certificate TypeCertificate of Analysis Date27/07/2020 Catalog NumberA2662
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Avidin, NeutrAvidin™, FITC conjugate > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.