missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ B-PER™ II Bacterial Protein Extraction Reagent (2X)

Efficiently lyse E.coli and other bacterial cells and gently extract native proteins with these nonionic detergent solutions.

Brand:  Thermo Scientific™ 78260

527.48 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10403815

  • 592.00 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

B-PER II Bacterial Protein Extraction Reagent (2X)
Liquid
250 mL
B-PER™
250 mL
Bacterial Protein Extraction Reagent
Store at room temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK402187 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number78260
Lot NumberZJ395946 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number78260
Lot NumberZF392033 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number78260
Lot NumberZC385945 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number78260
Lot NumberZA381559 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number78260
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ B-PER™ II Bacterial Protein Extraction Reagent (2X) > II Reagent (2X); 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.