Learn More
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals
Description
Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon

Specifications
Specifications
Color | Gray |
UN Number | UN1400 |
Quantity | 100 g |
Percent Purity | ≥99% |
Concentration or Composition (by Analyte or Components) | Sr ≤0.8% |
Odor | Odorless |
Weight | ≈7.3cm/100g |
Form | Rod |
Assay | (metals basis) |
Chemical Name or Material | Barium rod, 22mm (0.9 in.) dia. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
R05H113 | Certificate of Analysis | 05/12/2023 | 010103.22,010103.A1,010103.36 |
Q05I011 | Certificate of Analysis | 05/01/2023 | 010103.22,010103.A1,010103.36 |
P30H082 | Certificate of Analysis | 12/04/2022 | 010103.22,010103.A1,010103.36 |
S17H005 | Certificate of Analysis | 12/04/2022 | 010103.22,010103.A1,010103.36 |
D22U008 | Certificate of Analysis | 08/12/2021 | 010103.22,010103.A1,010103.36 |
Safety and Handling

- Barium rod
- Danger
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.