missing translation for 'onlineSavingsMsg'
Learn More

BD Cytofix™ Fixation Buffer

Can be used to fix unstained cells for subsequent immunofluorescent staining of intracellular cytokines. Can also be used to fix cells after immunofluorescent staining in order to preserve the light-scattering signals and fluorescent intensities of cells for analysis at a later time.

Brand:  BD 554655

211.11 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15817828

  • 455.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Intended to preserve human and rodent lymphoid cells for subsequent immunofluorescent staining of intracellular cytokines
  • Used to preserve light-scattering characteristics and fluorescence intensities of human and rodent hematopoietic cells that have been stained by immunofluorescence for subsequent flow cytometric analysis 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neutral pH Buffered Saline (PBS) with 4.21% Formaldehyde (w/w)
4°C
100 mL
Can be used to fix unstained cells for subsequent immunofluorescent staining of intracellular cytokines
Fixation Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Cytofix™ Fixation Buffer > Fixation Buffer; 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.