missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Human Interleukin-5 Flex Set Standard

Lyophilized human Interleukin

Brand:  BD 558463

115.56 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15859408

  • 116.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 25,000pg/ml (when reconstituted in 0.4mL Assay Diluent)
  • Regulatory status: RUO
  • Recombinant

Flow cytometry, controls

Order Info

100 tests.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
100 Tests
Flow cytometry
25000pg/mL
Human
Technical data sheet
Cytometric Ab Control Standards
RUO
4°C
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Cytometric Bead Array Human Interleukin-5 Flex Set Standard > Human interleukin-5 Flex set Standard

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.