Learn More
BD Human IL-1β Set II
An immunoassay design to fit individual laboratory needs
Brand:  BD 557953
980.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15859128
  • 985.00 € / Each
Explore more special offers
Includes: Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
Description
Description
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IL-1β in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3.9 to 250pg/mL
Human IL-1 beta Set II
-80°C
ELISA
20 Plates
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Human IL-1β Set II > Human IL-1β Set II
Research Use Only