missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil

Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.

Brand:  Bel-Art™ F13164-1419

 View more versions of this product

Product Code. 11392154

  • 250.00 € / Pack of 200
Estimated Shipment: 12-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.

  • Bright red polypropylene bags include black biohazard symbol
  • Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135°C (275°F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Made in USA
Specifications

Specifications

Biohazard Disposal Bag
Autoclavable
11 L
Zip Tie
Single-use
Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish
Non-sterile
1.50 mil
19 in.
14 in.
19 in.
14W x 19H in. (36 x 48cm)
2 to 4 gal.
ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact
Red
Used for biohazard waste
Polypropylene
0.279 mm
14 x 19 in.
48 cm
36 cm
48 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil > 14W x 19H in. (36 x 48cm); 2-4 gal.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.