missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Microcentrifuge Tube Beta Box

Protects from radioactive isotopes

Brand:  Bel-Art™ F24980-0015

Product Code. 15553835

This item is not returnable. View return policy

Description

Description

  • Made from 9.5 mm (0.8 in.) acrylic with tight-fitting joints
  • Crushed ice or water fits under rack
  • Holds 36 microcentrifuge tubes
Specifications

Specifications

Acrylic
Clear
Rectangular
36 x 1.5mL microcentrifuge tubes, water or crushed ice can be placed under rack
Non-Skid Feet, Tight Fitting Cover
Each
20 cm
6 cm
6 cm
Lid
Acrylic
Radioactive Isotope Protection
36 (1.5 mL Microcentrifuge Tubes)
Beta Box
8 x 3.5 x 2.3 in. (20.32 x 8.89 x 5.84 cm)
8.063 in.
3.5 in.
2.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Microcentrifuge Tube Beta Box > Acrylic Microcentrifuge Box

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.