missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical

Brand:  Fisher Chemical C/1500/60

73.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 10325220

  • 120.00 € / 1kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

1kg, Plastic powder jar

TRUSTED_SUSTAINABILITY
Specifications

Specifications

175°C
Beige to White
Solid
CaCl2·2H2O
147.02g/mol
10035-04-8
4.5 to 6.5
1 kg
149613
Plastic powder jar
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium chloride dihydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical > 1kg, Plastic powder jar

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.