Learn More
Certified Reference Material Fatty Acid Methyl Ester (FAME) Standard, Paragon Scientific™
Fatty Acid Methyl Ester (FAME) Certified Reference Material for the calibration and verification of laboratory instruments used for the determination of Fatty Acid Methyl Ester (FAME) content in middle distillates - Infrared spectrometry method, EN 14078
Brand:  Paragon Scientific CRM-FAGO
Product Code. 16020040
  • 290.00 € / 250mL
Explore more special offers
Description
Description
  • Certified Reference Material for Fatty Acid Methyl Ester (FAME) Content
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified via Round Robin
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Specifications
250 mL
UN 1202
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Certified Reference Material Fatty Acid Methyl Ester (FAME) Standard, Paragon Scientific™ > Quantity: 250mL
Nominal values quoted. Certified Values will vary from batch to batch.