Learn More
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Brand:  Kimble Chase™ 764500-0000
 View more versions of this product
Product Code. 13065803
  • 173.00 € / Pack of 100
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.5 μL
100/Pk.
32 mm
Borosilicate Glass
Capillary Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes > Capacity: 0.5uL; Length: 32mm; I.D.: 0.14mm; O.D.: 0.55mm