Learn More
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Brand:  Kimble Chase™ 7645000050
 View more versions of this product
Product Code. 13045813
  • 141.00 € / Pack of 100
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
100/Pk.
100 mm
Borosilicate Glass
Capillary Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes > Capacity: 50uL; Length: 100mm; I.D.: 0.79mm; O.D.: 1.09mm