missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
BIOPHARMAGREENER_CHOICE

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

Brand:  Thermo Scientific™ 475094

102.47 EUR valid until 2025-12-31
Use promo code "24284" to get your promotional price.


 View more versions of this product

Product Code. 10449672

  • 210.00 € / Pack of 60
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Alert:

Each lot is functionally tested for IgG binding.

This item is not returnable. View return policy

Description

Description

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
No
Without Lid
8 x 12
Clear
Polystyrene
96
Non-sterile
Round, Flat Bottom (F-well)
5
60/Cs.
Hydrophobic
Standard
Colorimetric
60 Cases
Microplate
400 μL
350 μL
PolySorp
60
None
Lipids
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number184191 Certificate TypeCertificate of Compliance Date17/03/2025 Catalog Number475094
Lot Number1282378 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog Number475094
Lot Number184536 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog Number475094
Lot Number133800 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number475094
Lot Number181375 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number475094
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates > Flat-Bottom, PolySorp, Pinchbar, 400μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.