Learn More

- Home
- Products
- Microplates
- Cell Culture Microplates
- Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Product Code. 10212811

Product Code. 10212811
Brand: Thermo Scientific™ 167008
Log in to see stock.

Use promo code "24107" to get your promotional price.
Explore more special offers
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available

Specifications
Specifications
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | With Lid |
Description | 96F well plate, Nunclon Delta, clear, with lid, Sterile (Pack of 1, Case of 50) |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 96 |
Sterility | Sterile |
Surface Treatment | Nunclon Delta |
Well Shape | Flat Bottom (F-well) |
No. per Case | 50 Microplates |
No. per Pack | 1 Microplate |
Quantity | 50/Cs. |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 10327461
-
Product Code. 10412741
-
Product Code. 10564581
-
Product Code. 10687551
-
Product Code. 10695951
-
Product Code. 15553115
-
Product Code. 15563115
-
Product Code. 10058820
-
Product Code. 10214171
-
Product Code. 10334791
-
Product Code. 10360691
-
Product Code. 10462012
-
Product Code. 10567131
-
Product Code. 10777241
-
Product Code. 10792552
-
Product Code. 15533115
-
Product Code. 15543115
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 10364131
340,00 € / Pack of 100 -
Catalog No. 10119831
302,00 € / Pack of 75 -
Catalog No. 10547781
253,00 € / Pack of 60 -
Catalog No. 11879181
88,80 € / Pack of 200 -
Catalog No. 10695951
195,00 € / Pack of 50 -
Catalog No. 10072151
361,00 € / Pack of 50 -
Catalog No. 11825275
59,45 € / Pack of 50 -
Catalog No. 10671722
Thermo Scientific ™ Nunc ™ 96-Well Polypropylene Sample Processing Thermo Scientific ™ Nunc ™ 96-Well Polypropylene Sample Processing
315,00 € / Pack of 120 -
Catalog No. 11512303
207,00 € / Pack of 500 -
Catalog No. 10344311
184,00 € / Pack of 50 -
Catalog No. 11548876
27,37 € / 100mL -
Catalog No. 11889610
29,90 € / Pack of 100 -
Catalog No. 10577391
299,52 € / Pack of 450 -
Catalog No. 10687551
200,00 € / Pack of 50
Documents
Scientific Resources
Brochures
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Lot Number185431 | Certificate TypeCertificate of Irradiation | Date06/04/2025 | Catalog Number167008 |
Lot Number185431 | Certificate TypeCertificate of Compliance | Date20/03/2025 | Catalog Number167008 |
Lot Number60904787 | Certificate TypeCertificate of Compliance | Date17/03/2025 | Catalog Number167008 |
Lot Number60903198 | Certificate TypeCertificate of Compliance | Date17/03/2025 | Catalog Number167008 |
Lot Number60900710 | Certificate TypeCertificate of Compliance | Date17/03/2025 | Catalog Number167008 |
- 1
Your input is important to us. Please complete this form to provide feedback related to the content on this product.