Learn More
Gibco™ Collagen I, Coated Plate, 24 well
Collagen I, Coated Plate, 24 well
Brand:  Gibco™ A1142802
Product Code. 12047556
  • 108.00 € / Pack of 5
Explore more special offers
Description
Description
Collagen I, Rat Tail is the cell culture workhorse of basic coating and gel preparations for growing attachment cells. Gibco™ Collagen I, Rat Tail is now available in pre-coated tissue culture plates to help save time at the bench. Designed to meet a number of different applications, our high quality, consistent Collagen I Coated Plates come in 3 formats – 96 well, 24 well and 6 well plates .
  • Ready to use, room temperature storage
  • Quality tested to help ensure consistency
  • Available in 3 different plate formats and in custom formats for a broad range of applications
    Easy to use, easy to store
    Collagen I, Coated Plates and lids are packed 5 per vacuum sealed foil pouch with a desiccant which can be stored at room temperature for a maximum shelf life of 2 years. Collagen I, rat tail is used to coat tissue culture treated plates. This is the same collagen material we sell in liquid form. The plates are ready to use with no further preparation required, simply seed your cells.
    Quality tested to help ensure consistency
    For maximum efficiency and consistency, Gibco™ Collagen I Coated Plates is designed to meet those expectations. We start with high quality rat tail collagen, the same material available in liquid form and follow a closely controlled, aseptic manufacturing process that produces a very consistent coating on the surface of tissue culture treated plates. We test each lot to help ensure we meet these standards.
    Available in different formats for a broad range of applications
    Extracellular matrix (ECM) proteins play an important role in the regulation of fundamental cellular processes and are widely used as a coating or matrix for tissue culture systems to permit the development of models that closely mimic in vivo conditions. Gibco™ Collagen I Coated Plates can be used in a broad range of applications, including the culturing of fresh or frozen hepatocytes (rat and human), as well as for neuronal, epithelial, and muscle primary cells. They are also ideal for culturing human corneal epithelial cells (HCEC) in defined media.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    5 plates are sealed in a foil pouch with a desicant for optimal shelf life. Store at room temperature.
    Gibco
    24
    Cell Culture Grade
    Room Temperature
    5/Pk.
    Culture Plate
    Cell Culture
    5 plates
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents Certificates
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number2809632 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberA1142803,A1142802,A1142801
    Lot Number2432893 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberA1142803,A1142802,A1142801
    Lot Number2298584 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberA1142803,A1142802,A1142801
    Lot Number2809633 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberA1142803,A1142802,A1142801
    Lot Number2432871 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberA1142803,A1142802,A1142801
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Gibco™ Collagen I, Coated Plate, 24 well > 5 plates