missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces

Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.

Brand:  Thermo Scientific™ 436017

894.25 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10133682

  • 941.32 € / Pack of 15
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.

Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)

  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
Colorimetric
Affinity
Microplate
120 μL
50 μL
Streptavidin
Square, Flat Bottom (F-well)
1
15/Cs.
Without Lid
15 Cases
Clear
Polystyrene
384
Non-sterile
120 μL
15
Unspecified
Biotinylated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number165176 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436017
Lot Number166287 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436017
Lot Number178088 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436017
Lot Number166285 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436017
Lot Number179473 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436017
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.