Learn More
Nabertherm™ Compact Tube Furnace with C550 Controller
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
4995.00 € - 12960.00 €
Specifications
Controller Type C550
Type Compact Tube Furnace
View More Specs
Products 10
Product Code Brand Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Price Quantity & Availability  
17338079
View Documents
Nabertherm™
R-152O2AN
50 mm 250 mm 1200°C
4995.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17348079
View Documents
Nabertherm™
R-152O3AN
50 mm 250 mm 1300°C
5290.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17378079
View Documents
Nabertherm™
R-252O2AN
50 mm 500 mm 1200°C
5570.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17388079
View Documents
Nabertherm™
R-252O3AN
50 mm 500 mm 1300°C
6080.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17338089
View Documents
Nabertherm™
R-322O2AN
120 mm 500 mm 1200°C
6745.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17348089
View Documents
Nabertherm™
R-322O3AN
120 mm 500 mm 1300°C
7555.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17398089
View Documents
Nabertherm™
R-472O2AN
170 mm 750 mm 1200°C
9830.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17308099
View Documents
Nabertherm™
R-472O3AN
170 mm 750 mm 1300°C
11020.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17358099
View Documents
Nabertherm™
R-572O2AN
170 mm 1000 mm 1200°C
11460.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
17368099
View Documents
Nabertherm™
R-572O3AN
170 mm 1000 mm 1300°C
12960.00 €
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
Description
Description
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Five programs and four freely programmable ramps, controller C 550 with 10 freely adjustable programs and 20 segments
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Specifications
C550
Compact Tube Furnace
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Nabertherm™ Compact Tube Furnace with C550 Controller