Learn More
Fisherbrand™ Connecting Cable
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
40.55 € - 44.70 €
Specifications
Type Connecting Cable
View More Specs
Products 3
Product Code Brand Connector Types Price Quantity & Availability  
Product Code Brand Connector Types Price Quantity & Availability  
11766348
View Documents
Fisherbrand™
9001041-003U
Banana
42.40 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
11776398
View Documents
Fisherbrand™
9001041-010B
BNC
44.70 €
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
11736348
View Documents
Fisherbrand™
9001041-003I
DIN
40.55 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Connecting Cable with BNC Plug
  • For combined ion-sensitive electrode with solid membranes
  • For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
Specifications
Connecting Cable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Connecting Cable