Learn More
Fisherbrand™ Connecting Cable
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
Brand:  Fisherbrand™ 9001041-003I
 View more versions of this product
Product Code. 11736348
  • 40.55 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Connecting Cable with DIN Plug
  • For combined ion-sensitive electrode with solid membranes
  • For use with measuring agriculture, water analysis, and calibration of ionic electrodes
  • For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug
Specifications
Specifications
DIN
For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug.
1 m
Connecting Cable
0°C to 50°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Connecting Cable > Connector Type: DIN