Learn More
Gilson™ Connection Kit
Brand:  Gilson™ F1825054
 View more versions of this product
Product Code. 10113002
  • 248.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Connector kit accessories for the MINIPULS™ 3 and Evolution peristaltic pumps come in a variety of types: metal sleeve connectors for small bore tubing, connector kits for flanged tubing, or plastic connectors for large bore tubing.
  • These kits contain five connectors each.
  • The connectors are available in a three sizes to fit a variety of tubing of different inner diameters.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 mm
1.0 to 1.65 mm
5 Set
Connection Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ Connection Kit > O.D.: 3mm