Learn More
Contec™ SATWIPES™ Presaturated Wipes



Prepare for a variety of cleanrooms. The SATWIPES™Presaturated Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
Brand: Contec™ SAT-C1-100
300.14 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
- Durable, presaturated wipes with a choice of solvent saturations on perforated rolls
- Polyester Nonwoven are ideal for critical applications in ISO Class 5 environments
- SATWIPES with Polyester/Cellulose Nonwoven Wipes offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
- Designed for general-purpose cleaning and surface preparation and for wiping process tools and other equipment and for use prior to cleanroom entry
Order Info
Packaged one hundred 6 x 9 in. (15 x 23cm) wipers on a perforated roll. Most SATWipes cases contain 12 presaturated rolls plus one canister; 19-150-0711 and -0712 packed 18 refills per case without a canister. Custom-blend substrates and additional wiper sizes available through your Fisher Safety Customer Service Representative. Empty canisters for wipes is available: 19-171-955.

Specifications
Pre-saturated Wipe | |
23 cm | |
15.2 cm | |
Not Autoclavable | |
23 x 15 cm (9 x 6 in.) | |
12 Pk. | |
100% IPA |
9 in. | |
6 in. | |
Polyester Cellulose | |
Single-use | |
Excellent wipes for less critical applications, compatible with ISO class 6 environments and wipes down prior to entry into cleanroom | |
1-ply | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- X1200 SATWipes cellulose/polyester wipes, 15x23cm,
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
945193-0-0-1 | Certificate of Quality | 06/01/2023 | SAT-C1-100 |
881031-0-0-1 | Certificate of Compliance | 26/05/2022 | SAT-C1-100 |
881030-0-0-1 | Certificate of Compliance | 04/03/2022 | SAT-C1-100 |
878887-0-0-1 | Certificate of Compliance | 26/11/2021 | SAT-C1-100 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.