Learn More
Corning™ Thermowell™ GOLD Polypropylene PCR Tubes
GREENER_CHOICE
Designed for precise fit in heat blocks
Brand:  Corning™ 3745
 View more versions of this product
Product Code. 10092122
  • 130.99 € / Pack of 1000
Explore more special offers
Description
Description
Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase-free
Clear
500/Pk.
Heat blocks to optimize heat transfer
Non-sterile
10,000 x g
Autoclavable
Thin Wall PCR Tube
Clear
0.2 mL
Polypropylene
Flat
Polypropylene
Flat Cap
Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1420162 Certificate TypeCertificate of Processing Date30/01/2025 Catalog Number3745
Lot Number1418094 Certificate TypeCertificate of Processing Date18/12/2024 Catalog Number3745
Lot Number1418095 Certificate TypeCertificate of Processing Date17/12/2024 Catalog Number3745
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Thermowell™ GOLD Polypropylene PCR Tubes > Clear; Thermowell GOLD; Flat; 0.2 mL