Learn More
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
Brand: Corning™ 8162
267.50 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
- Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Specifications
Cellulose Acetate | |
Sterile | |
0.5 mL |
0.45 μm | |
Spin-X |
Product Suggestions
Customers who viewed this item also viewed