Learn More
R&D Systems™ Cultrex 3D Culture BME Cell Invasion Assay, 96 well
A three-dimensional, high content format for quantitating the degree to which invasive cells penetrate basement membrane components, in vitro, in response to chemoattractants and/or inhibiting compounds.
Brand: R&D Systems™ 3500-096-K
Includes: 10X Spheroid Formation ECM, 600uL; 3D Culture Qualified 96 Well Spheroid Formation Plate; Invasion Matrix, 6mL
Description
Basement membranes are continuous sheets of specialized extracellular matrix that form an interface between endothelial, epithelial, muscle, or neuronal cells and their adjacent stroma. They not only support cells and cell layers, but they also play an essential role in tissue organization that influences cell adhesion, migration, proliferation, and differentiation. Basement membranes are degraded and regenerated during development and wound repair, and they are major barriers to invasion by metastatic tumor cells.
Provides a more physiological approach for assessing tumor invasion and offers a visual component
Kit matrix of basement membrane proteins forms a hydrogel network on which invasive cells can travel
At this point, invasion modulating agents can be applied to the system to evaluate the impact on cell response. Cell invasion is visualized microscopically and can be quantified through image analysis software.
- Cultrex Spheroid Formation Matrix
- 3-D Culture Qualified 96 Well Spheroid Formation Plate with Strip Seals
- Cultrex Spheroid Invasion Matrix
Specifications
The unopened product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Each | |
Mouse |
Product Suggestions
Customers who viewed this item also viewed
For research use only. Not for diagnostic use.