missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Methionine, 99% Product Code.: 11464717

Thermo Scientific Chemicals D-Methionine, 99%

Product Code. 11464717
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11464717

Brand: Thermo Scientific Chemicals B21213.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.

Solubility
Soluble in water. 53 g/L (20°C).

Notes
Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-67-4
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00002622
InChI Key FFEARJCKVFRZRR-SCSAIBSYSA-N
Synonym d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acidShow MoreShow Less
PubChem CID 84815
ChEBI CHEBI:16867
IUPAC Name (2R)-2-amino-4-methylsulfanylbutanoic acid
SMILES CSCC[C@@H](N)C(O)=O

Specifications

Melting Point ∼280°C (decomposition)
Beilstein 1722293
Merck Index 14,5975
Solubility Information Soluble in water. 53g/L (20°C).
Quantity 5 g
Formula Weight 149.21
Optical Rotation −23° (c=2 in 2N HCl)
Percent Purity 99%
Chemical Name or Material D-Methionine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ13I005 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog NumberB21213.14,B21213.22,B21213.06
Lot NumberU29I013 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog NumberB21213.14,B21213.22,B21213.06
Lot Number10164372 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21213.14,B21213.22,B21213.06
Lot Number10026654 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21213.14,B21213.22,B21213.06
Lot Number10126499 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21213.14,B21213.22,B21213.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Methionine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.