missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Tyrosine, 98% Product Code.: 10616344

Thermo Scientific Chemicals D-Tyrosine, 98%

Product Code. 10616344
10 g, Glass Bottle
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10616344

Brand: Thermo Scientific Chemicals 226910100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-02-5
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063073
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosineShow MoreShow Less
PubChem CID 71098
ChEBI CHEBI:28479
IUPAC Name (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
Residue after Ignition 0.1% max.
Beilstein 14, 605
Specific Rotation +10° to +12° (20°C, 589nm) (c=5, 1N HCl) (after 2 hrs.)
Merck Index 14, 9839
Specific Rotation Condition +11.30° (20°C c=5,1N HCl)
Quantity 10 g
Formula Weight 181.19
Percent Purity ≥97.5%
Physical Form Powder or Crystals
Chemical Name or Material D-Tyrosine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467335 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number226910025,226910100
Lot NumberA0400489 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number226910025,226910100
compliance-icons
Product Identifier
  • D-Tyrosine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Tyrosine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.