Learn More
Dexamethasone 9,11-epoxide, MedChemExpress
MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Brand:  Medchem Express HY-N0348
Encompass_Preferred
 View more versions of this product
Product Code. 16489056
  • 201.15 € / 100mg
Explore more special offers
Description
Description
Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Specifications
Specifications
Dexamethasone 9,11-epoxide
White
C22H28O5
DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
372.45
99.3%
Solid
24916-90-3
Research
100 mg
C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O
372.45
Research
COVID-19-immunoregulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Dexamethasone 9,11-epoxide, MedChemExpress