missing translation for 'onlineSavingsMsg'
Learn More
Dichlorobis(tricyclohexylphosphine)palladium(II), Thermo Scientific Chemicals Product Code.: 15406625

Dichlorobis(tricyclohexylphosphine)palladium(II), Thermo Scientific Chemicals

Product Code. 15406625
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15406625

Brand: Thermo Scientific Alfa Aesar 044844.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

It is used as catalyst for C-C and C-N coupling reaction. It is used in the homogeneous carbonylation of chloroarenes under mild conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29934-17-6
Molecular Formula C36H66Cl2P2Pd
MDL Number MFCD00191830
Quantity 1 g
Synonym dichlorobis tricyclohexylphosphine palladium ii, bis tricyclohexylphosphine palladium ii dichloride, dichlorobis tricyclohexylphosphine palladium, bis tricyclohexylphosphine-palladium ii dichloride, bis tricyclohexylphosphine palladium ii-dichloride, bis tricyclo-hexylphosphine palladium ii dichloride, bis tricyclohexylphosphine palladium ii chloride
Solubility Information Moderately soluble in dichloromethane and chloroform. Sparingly soluble in benzene and toluene. Insoluble in water,alcohols,acetone,diethyl ether,and hexanes.
InChI Key VUYVXCJTTQJVKJ-UHFFFAOYSA-L
SMILES Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
IUPAC Name dichloropalladium;tricyclohexylphosphane
Molecular Weight (g/mol) 738.19
PubChem CID 11050900
Formula Weight 738.18
Concentration or Composition (by Analyte or Components) Pd 14.4%
Physical Form Powder
Chemical Name or Material Dichlorobis (tricyclohexylphosphine)palladium (II)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW10E026 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044844.03
Lot NumberM25C001 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044844.03
Lot NumberY19G037 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044844.03
Lot NumberQ05H024 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044844.03
Lot NumberY13F041 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044844.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichlorobis(tricyclohexylphosphine)palladium(II), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.