missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)
Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Brand: BD 236920
This item is not returnable.
View return policy
Description
Applications:
- Used with or without blood for cultivation and isolation of variety of organisms
- Used for sterility testing
Certifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP

Specifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Dehydrated | |
Agar | |
Isolation and cultivation of non-fastidious and fastidious microorganisms | |
2 kg |
2°C to 30°C | |
Tryptic Soy Agar | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Pail |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) > Tryptic Soy Agar (Soybean-Casein Digest Agar Medium); Quantity: 2kg
Spot an opportunity for improvement?Share a Content Correction