Learn More
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)
Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Brand:  BD 236950
 View more versions of this product
Product Code. 11768822
  • 170.00 € / 500g
Explore more special offers
Description
Description
Applications:
  • Used with or without blood for cultivation and isolation of variety of organisms
  • Used for sterility testing
Certifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Dehydrated
Agar
Isolation and cultivation of non-fastidious and fastidious microorganisms
500 g
2°C to 30°C
Tryptic Soy Agar
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L
Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) > Tryptic Soy Agar (Soybean-Casein Digest Agar Medium); 500g