missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Horizontal Large Gel Systems: Microplate Combs
For large gel systems
Brand: Fisherbrand™ A2-MT2D
This item is not returnable.
View return policy
Description

Specifications
Owl A2 and A5 Large Gel Electrophoresis System | |
Microplate | |
0.15 cm | |
0.09 in. |
Micro Well Comb | |
42 | |
20 μL | |
0.25 cm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Horizontal Large Gel Systems: Microplate Combs > Comb; 42-tooth; 1.5mm thick; Microplate format; For horizontal large gel systems
Spot an opportunity for improvement?Share a Content Correction