missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Wide Neck Round Bottom Boiling Flask

Made of borosilicate glass

Brand:  Pyrex 1120/29D

 View more versions of this product

Product Code. 11991718

This item is not returnable. View return policy

Description

Description

  • Complies with ISO 24450 and DIN 12353
  • With wide neck to facilitate addition of solid samples etc
  • Resistant to chemical corrosion and thermal shock
TRUSTED_SUSTAINABILITY
Specifications

Specifications

80 mm
Borosilicate Glass
10
Bulb: 206 mm
6 L
355 mm
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex™ Wide Neck Round Bottom Boiling Flask > Capacity: 6000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.