Learn More
S Murray and Co™ Dissecting Set
Use this set for a variety of applications. S Murray™ Dissecting Set is fully guaranteed in normal use and subject to stringent quality inspection procedures.
Brand:  S Murray and Co™ Y261/72
Product Code. 12774416
  • 178.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Unless otherwise stated instruments are made from stainless steel.
  • 25 piece set
  • Includes six awls
Specifications
Specifications
Plastic
1 Forceps, 125mm, straight, fine points; 5 Scalpel blades; 1 Forceps, 125mm, straight, blunt tips; 5 Scalpel blades; 2 Needles, Stainless Steel in metal handle; 2 Scalpel blade holders, nickel plated; 2 Seekers, Stainless Steel in metal handle; 1 Scissors, curved, fine points, 6 Dissectingawls, Stainless Steel in metal handle
Stainless Steel and Metal
Dissecting Set
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
S Murray and Co™ Dissecting Set > Stainless Steel and Metal