missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Leucine, 99+% Product Code.: 10741641

Thermo Scientific Chemicals DL-Leucine, 99+%

Product Code. 10741641
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
250 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10741641

Brand: Thermo Scientific Chemicals 172130250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-39-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063087
InChI Key ROHFNLRQFUQHCH-UHFFFAOYNA-N
Synonym dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,dShow MoreShow Less
PubChem CID 857
ChEBI CHEBI:25017
IUPAC Name 2-amino-4-methylpentanoic acid
SMILES CC(C)CC(N)C(O)=O

Specifications

Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.3% max.
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH2CH(NH2)CO2H
Beilstein 04, 447
Merck Index 14, 5451
Sulfated Ash 0.1% max.
Quantity 25 g
Formula Weight 131.17
Percent Purity 99.0 to 101.0% (on dry substance)
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Leucine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461526 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number172132500,172130250
Lot NumberA0420300 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number172132500,172130250
Lot NumberA0405405 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number172132500,172130250
Lot NumberA0441179 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number172132500,172130250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Leucine, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.