missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Proline, 99% Product Code.: 10733561

Thermo Scientific Chemicals DL-Proline, 99%

Product Code. 10733561
25 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10733561

Brand: Thermo Scientific Chemicals 174030250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-36-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00005250
InChI Key ONIBWKKTOPOVIA-UHFFFAOYSA-N
Synonym dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
PubChem CID 614
ChEBI CHEBI:26271
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Melting Point 208.0°C to 210.0°C
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Residue after Ignition 0.1% max.
Beilstein 22, 4
Fieser 09,393
Merck Index 15, 7895
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol, sparingly soluble in chloroform, benzene and, acetone, insoluble in ether
Water 0.5% max. (K.F.)
Quantity 25 g
Formula Weight 115.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-Proline, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453381 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number174030250,174030050
Lot NumberA0434997 Certificate TypeCertificate of Analysis Date05/10/2021 Catalog Number174030250,174030050
Lot NumberA0390144 Certificate TypeCertificate of Analysis Date08/05/2020 Catalog Number174030250,174030050
Product Identifier
  • DL-Proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Proline, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.