Learn More
Gibco™ DMEM, high glucose, no glutamine, no phenol red

DMEM, high glucose, no glutamine, no phenol red
Brand: Gibco™ 31053044
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.DMEM is unique from other media as it contains 4 times the concentrations of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Product Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM product made in our Grand Island facility (31053-036). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
ISO 13485 standards, FDA | |
DMEM Medium, 500mL, Liquid Form, With High Glucose, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture | |
DMEM Medium | |
No Glutamine, No Phenol Red, No Sodium Pyruvate, No HEPES | |
10 x 500 mL |
2 to 8°C | |
Liquid | |
High Glucose | |
Mammalian cell culture | |
For in-vitro diagnostic use |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.