Learn More
Justrite™ Drum Cabinet Vertical Single Drum Storage
Brand:  Justrite™ 8962001
Product Code. 12933425
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
2 door, manual, with removable, adjustable shelf, steel can accommodate pump or funnel yellow 1651mm x 864mm x 864mm (w x h x d) 1 x 200L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Justrite™ Drum Cabinet Vertical Single Drum Storage >