missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ mRNA DIRECT™ Purification Kit

The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues.

Marca:  Invitrogen™ 61012

1105.80 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.



 vedi altre versioni di questo prodotto

Codice prodotto. 10563755

  • 1224.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Esplora altre offerte speciali

Includes: 10mL Dynabeads oligo (dT)25, 60mL Lysis/Binding Buffer, 120mL Washing Buffer A, 60mL Washing Buffer B, 15mL 10mM Tris-HCl (Elution buffer), Contains enough reagents for 40 standard isolations.
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues. The isolated mRNA is suitable for use in all downstream applications.

Advantages of the Dynabeads mRNA DIRECT Kit:
Fast—15 minute procedure yields pure, intact mRNA
Highly pure mRNA isolation—best choice upstream of cDNA synthesis
Sensitive mRNA isolation—enables cDNA synthesis and cDNA library construction from ultra-small starting samples (enables cDNA library construction from a single cell)

System overview
The isolation protocol relies on base pairing between the polyA residues at the 3' end of most mRNA, and the oligo (dT)25 residues covalently coupled to the surface of the Dynabeads. Other RNA species lacking a polyA tail will not hybridize to the beads and are readily washed away. Ribosomal RNA, DNA, proteins, and small RNA molecules (such as transfer RNA, micro RNA, and small nucleolar RNA) do not bind to the beads and are discarded. RNase inhibiting agents in the Lysis/Binding Buffer together with stringent hybridization and washing conditions ensure the isolation of pure, intact mRNA from crude samples rich in RNase, without the use of strong chaotropic agents. The mRNA purification beads specifically target and capture the mRNA transcriptome from an extremely wide variety of crude starting samples (see protocol). 1 mg of Dynabeads Oligo (dT)25 beads (200 μL) binds up to 2 μg of mRNA. A typical mammalian cell contains about 10–30 pg of total RNA, from which 1–5% is mRNA.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifica

Specifica

• 10 mL Dynabeads Oligo (dt)25, 4°C
• 60 mL Lysis/Binding Buffer; 4°C
• 120 mL Washing Buffer A; 4°C
• 60 mL Washing Buffer B; 4°C
• 15 mL 10 mM Tris-HCl; 4°C

10 to 100 μL
mRNA
Automated Liquid Handling Systems
Beads may be reused for multiple extractions
Room Temperature
2 μg mRNA per 200 μL of beads (Binding capacity)
Magnetic Bead
Cells, Plants, Plasma, Serum, Tissue, Total RNA, Blood
High-throughput Compatible
RT-PCR, qPCR, cDNA library construction, microarray analysis
10 mL
Cells: ≤106
Plant: ≤400 mg
Tissue: ≤200 mg
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
Sicurezza e movimentazione
Show More
SDS
Documenti

Documenti

Certificati

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3043147 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number61012
Lot Number3149114 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number61012
Lot Number2793369 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number61012
Lot Number2840184 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number61012
Lot Number3041731 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number61012
Offerte speciali

Offerte speciali

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Invitrogen™ Dynabeads™ mRNA DIRECT™ Purification Kit > 10 mL

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato

For Research Use Only. Not for use in diagnostic procedures.