Learn More
Terumo™ Luer-Lock Tip Three-Part Syringe
Terumo™ Luer-Lock Tip Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
Brand:  Terumo™ 8SS20L1
 View more versions of this product
Product Code. 11866071
  • 72.20 € / Pack of 50
Edge
Explore more special offers
Description
Description
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Luer-Lock Three-Part Syringes feature a unique double beveled design for greater patient comfort.
  • Silicone coated barrel for a smooth plunger action
  • Clear barrel to allow quick recognition of air bubbles
  • Easy to read markings
  • Latex free
  • Sterile and packed in individually wrapped tamper peel packs
  • Syringes are available with luer lock, luer slip, catheter and eccentric tips
  • Available in a wide range of sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Polypropylene
For the injection of fluids immediately after filling into the human body, for aspiration of fluids or for the sampling of blood or other body fluids
Luer-Lock Tip Three-Part Syringe, Concentric
50 Pk.
Luer Lock
Sterile
Class IIa, sterile product following MDD 93/42/EEC, CE 0197
Screen printed graduation
Yes
Single-use
20 mL
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Terumo™ Luer-Lock Tip Three-Part Syringe > Volume: 20 mL