Learn More
Eppendorf™ epT.I.P.S.™ Standard Pipette Tips
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
45.45 € - 92.77 €
Specifications
Autoclavable | Autoclavable |
---|---|
Compatibility | Universal |
Format | Bulk |
Tip Style | Universal |
Product Type | Pipette Tips |
Product Code | Brand | Volume (Metric) | Length (Metric) | Color | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Length (Metric) | Color | Packaging | Price | Quantity & Availability | |||
10708972
![]() |
Eppendorf™
0030000854 |
0.5 to 20 μL | 46 mm | Light Gray | 1000 Tips (2 Bags x 500 Tips) |
68.60 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10157291
![]() |
Eppendorf™
0030000978 |
100 to 5000 μL | 120 mm | Purple | 500 Tips (5 Bags x 100 Tips) |
63.40 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
10333682
![]() |
Eppendorf™
0030000765 |
1000 to 10,000 μL | 165 mm | Turquoise | 200 Tips (2 Bags x 100 Tips) |
65.06 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
10199171
![]() |
Eppendorf™
0030000781 |
1000 to 10,000 μL | 243 mm | Turquoise | 200 Tips (2 Bags x 100 Tips) |
92.77 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
11920537
![]() |
Eppendorf™
0030000889 |
2 to 200 μL | 53 mm | Clear | 1000 Tips (2 Bags x 500 Tips) |
48.24 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10248621
![]() |
Eppendorf™
0030000870 |
2 to 200 μL | 53 mm | Yellow | 1000 Tips (2 Bags x 500 Tips) |
46.38 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10177241
![]() |
Eppendorf™
0030000897 |
20 to 300 μL | 55 mm | Orange | 1000 Tips (2 Bags x 500 Tips) |
46.38 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10370842
![]() |
Eppendorf™
0030000919 |
50 to 1000 μL | 71 mm | Blue | 1000 Tips (2 Bags x 500 Tips) |
46.38 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10772559
![]() |
Eppendorf™
0030000927 |
50 to 1000 μL | 71 mm | Clear | 1000 Tips (2 Bags x 500 Tips) |
46.98 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10218671
![]() |
Eppendorf™
0030000951 |
250 to 2500 μL | 115 mm | Red | 500 Tips (5 Bags x 100 Tips) |
54.32 €
Pack of 500 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||
10197241
![]() |
Eppendorf™
0030000935 |
50 to 1250 μL | 76 mm | Green | 1000 Tips (2 Bags x 500 Tips) |
75.65 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10788962
![]() |
Eppendorf™
0030000811 |
0.1 to 10 μL | 34 mm | Dark Gray | 1000 Tips (2 Bags x 500 Tips) |
75.20 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10426461
![]() |
Eppendorf™
0030000838 |
0.1 to 20 μL | 40 mm | Medium Gray | 1000 Tips (2 Bags x 500 Tips) |
78.21 €
Pack of 1000 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||
10744587
![]() |
Eppendorf™
0030000730 |
50 to 1250 μL | 103 mm | Dark Green | 1000 Tips (4 Bags x 250 Tips) |
76.75 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
10541159
![]() |
Eppendorf™
0030000900 |
20 to 300 μL | 55 mm | Orange | 1000 Tips (2 Bags x 500 Tips) |
45.45 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||
Description
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
- Minimal attachment and ejection forces with the highest level of tightness
- The ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
- Fine graduations make visual checks of the pipetting volume even easier
- Fine retracted or extended tips simplify work with small volumes
- Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
Specifications
Autoclavable | |
Bulk | |
Pipette Tips | |
Yes | |
Fine |
Universal | |
Universal | |
Eppendorf Pipetter | |
epT.I.P.S. |