missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Specifications
Specifications
Packaging | Metal can |
Quantity | 5 L |
Molecular Formula | C2H6O |
Physical Form | Liquid |
Color | Colorless |
Melting Point | -114°C |
Boiling Point | 78°C |
Formula Weight | 46.07g/mol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527610 | Certificate of Analysis | 18/02/2025 | E/0550DF/25,E/0550DF/C17,E/0550DF/21,E/0550/08,E/0550DF/17,E/0550/17,E/0550/PC17 |
2525705 | Certificate of Analysis | 05/02/2025 | E/0550DF/25,E/0550DF/15,E/0550DF/C17,E/0550DF/21,E/0550/08,E/0550DF/17,E/0550/17,E/0550/PC17 |
Safety and Handling


Product Identifier
- Ethanol (95% v/v)
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction